missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRB3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22190-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
PRB3 protein is identical to the gross cystic disease fluid protein GCDFP-15, secretory actin binding protein (SABP) and glycoprotein 17 (GP-17). GCDFP-15 is expressed by breast carcinomas (primary and metastatic lesions) with apocrine features.Spécification
PRB3 | |
Polyclonal | |
Unconjugated | |
PRB3 | |
G1, PRB3, PRG, Proline rich protein G1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5544, 639802 | |
-20°C | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q04118 | |
PRB3, Prb3-ps | |
PRB3 Fusion Protein Ag16760 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |