missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Precoated TLC and PLC Glass Plates
Average particle size is 15μm with a 5-20μm distribution
2110.00€
Code produit | Marque | Dimensions (L x l) | Matériau | Type | Quantité | Prix | Quantité et disponibilité | ||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Dimensions (L x l) | Matériau | Type | Quantité | Prix | Quantité et disponibilité | ||
17869671
|
MilliporeSigma™
1.15389.0001 |
20 x 20 cm | Gel de silice 60 RP-18 F254 s | Plaque pour CCM en silice modifiée RP | 25/paquet |
2110.00€
25 pièces |
Expédition estimée: 06-06-2024
Connectez-vous pour voir le stock disponible |
||
Description
- Choose from several media to match your separation requirements: silica gel 60 (with and without F254 fluorescent indicator), chemically modified silica gel (reversed phases RP-2, RP-8 and RP-18), aluminum oxide 60 with F254 indicator, cellulose, and cellulose PEI
- 60Å silica gel is suitable for all common gel separations including weak to strong polar analytes
- Silica can also be used for applications requiring charring procedures
- Aluminum oxide or alumina plates are ideal for separating hormones, alkaloids and antibiotics
- For partition chromatography, select cellulose plates; and for separation of weak anion exchangers, including peptide and amino acids, use cellulose PEI plates
Certifications
ISO 9001:2000
†With acid-stable fluorescent indicator.
‡PEI (polyethyleneimine) should be stored at 4°C (39°F). If stored at room temperature, PEI plates may turn slightly yellow, which could affect separation. To minimize yellow color, predevelop in distilled water.
Spécification
Verre |