missing translation for 'onlineSavingsMsg'
Learn More

PRPF3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 10106-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16836273

  • 325.38€ / 150µL
Expédition estimée: 06-10-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody is a rabbit polyclonal antibody raised against an internal region of human PRPF3. A predicted molecular mass of 77 kDa and the gene is named PRPF3 because it is homologous to the yeast PRP3 gene. We noticed that although the predicted molecu

The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs. The removal of introns from nuclear pre-mRNAs occurs on complexes called spliceosomes, which are made up of 4 small nuclear ribonucleoprotein (snRNP) particles and an undefined number of transiently associated splicing factors. PRPF3 is 1 of several proteins that associate with U4 and U6 snRNPs.
Spécification

Spécification

PRPF3
Polyclonal
Unconjugated
PRPF3
HPRP3, HPRP3P, Pre mRNA splicing factor 3, PRP3, Prp3p, PRPF3, RP18, U4/U6 snRNP 90 kDa protein
Rabbit
Antigen Affinity Chromatography
RUO
361995, 70767, 9129
-20°C
Liquid
Western Blot, Immunoprecipitation
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43395, Q922U1
Prpf3
PRPF3 Fusion Protein Ag0154
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions