missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSG11 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16352-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
PSG11, also named as PSG13, PSG14, PSBG-11 and PSBG-13, belongs to the immunoglobulin superfamily and CEA family. PSBG are produced in high quantity during pregcy. PSG11 and other PSGs induce dose-dependent secretion of anti-inflammatory cytokines by human monocytes.Spécification
PSG11 | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UQ72 | |
PS beta G 11, PS beta G 13, PSBG 11, PSBG 13, PSG11, PSG13, PSG14 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
5680 |
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
PSG11 | |
Liquid | |
PSG11 | |
PSG11 Fusion Protein Ag9562 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |