missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMB3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15983-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The proteasome is a multicatalytic proteinase complex with a highly ordered ring-shaped 20S core structure. The core structure is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a member of the proteasome B-type family, also known as the T1B family, that is a 20S core beta subunit. Pseudogenes have been identified on chromosomes 2 and 12.Spécification
PSMB3 | |
Polyclonal | |
Unconjugated | |
PSMB3 | |
20S proteasome, HC10 II, Proteasome chain 13, Proteasome component C10 II, Proteasome subunit beta type 3, Proteasome theta chain, PSMB3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26446, 29676, 5691 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P40112, P49720, Q9R1P1 | |
PSMB3 | |
PSMB3 Fusion Protein Ag8784 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |