missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24142-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a WASP interacting protein (WIP)-related protein. It has been shown that this protein has a role in the WASP-mediated organization of the actin cytoskeleton and that this protein is a potential link between the activated platelet-derived growth factor receptor and the actin polymerization machinery.Spécification
PSMC3 | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P17980 | |
Proteasome subunit P50, PSMC3, Tat binding protein 1, TBP 1, TBP1 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
5702 |
Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Unconjugated | |
PSMC3 | |
Liquid | |
PSMC3 | |
PSMC3 Fusion Protein Ag21408 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |