Learn More
Lenz Laborglasinstrumente™ Agitateur en PTFE
Ideal for routine stirring and mixing applications. Lenz Laborglasinstrumente™ Stirrers with PTFE Blade and Journal feature a shaft manufactured from durable DURAN™ tubing.
Marque: Lenz Laborglasinstrumente™ B3972565
Informations supplémentaires : Poids : 0.07200kg
Description
- Precision calibrated glass tube
- Ground and polished bearing surface
- With snap fastener for replacement of PTFE blades