Learn More
Lenz Laborglasinstrumente™ Agitateur en PTFE
Ideal for routine stirring and mixing applications. Lenz Laborglasinstrumente™ Stirrers with PTFE Blade and Journal feature a shaft manufactured from durable DURAN™ tubing. 
Marque:  Lenz Laborglasinstrumente™ B3972573
Informations supplémentaires : Poids : 0.08100kg
Code produit. 10000440
  • 119.00€ / 1 pièce
Expédition estimée: 22-05-2024
Description
  • Precision calibrated glass tube
  • Ground and polished bearing surface
  • With snap fastener for replacement of PTFE blades