missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTPN13 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25944-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a member of the protein tyrosine phosphatase (PTP) family. PTPs are signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP is a large intracellular protein. It has a catalytic PTP domain at its C-terminus and two major structural domains: a region with five PDZ domains and a FERM domain that binds to plasma membrane and cytoskeletal elements. This PTP was found to interact with, and dephosphorylate, Fas receptor and IkappaBalpha through the PDZ domains. This suggests it has a role in Fas mediated programmed cell death. This PTP was also shown to interact with GTPase-activating protein, and thus may function as a regulator of Rho signaling pathways. Four alternatively spliced transcript variants, which encode distinct proteins, have been reported.Spécification
PTPN13 | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q12923 | |
FAP 1, hPTPE1, PNP1, PTP BAS, PTP BL, PTP E1, PTP1E, PTPL1, PTPLE, PTPN13 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
5783 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
PTPN13 | |
Liquid | |
PTPN13 | |
PTPN13 Fusion Protein Ag22854 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |