missing translation for 'onlineSavingsMsg'
Learn More

PTPRE Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13922-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802354

  • 329.85€ / 150µL

Date d’expédition estimée: 18-06-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene is a member of the protein tyrosine phosphatase family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. Two alternatively spliced transcript variants of this gene have been reported, one of which encodes a receptor-type PTP that possesses a short extracellular domain, a single transmembrane region, and two tandem intracytoplasmic catalytic domains; Another one encodes a PTP that contains a distinct hydrophilic N-terminus, and thus represents a nonreceptor-type isoform of this PTP. Studies of the similar gene in mice suggested the regulatory roles of this PTP in RAS related signal transduction pathways, cytokines induced SATA signaling, as well as the activation of voltage-gated K+ channels.
Spécification

Spécification

PTPRE
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23469
HPTPE, PTPE, PTPRE, R PTP EPSILON
Rabbit
IgG
150 μL
-20°C
Primary
5791
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTPRE
Liquid
PTPRE
PTPRE Fusion Protein Ag4932
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Vidéos
FDS
Documents