missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTRH2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 51006-2-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a mitochondrial protein with two putative domains, an N-terminal mitochondrial localization sequence, and a UPF0099 domain. In vitro assays suggest that this protein possesses peptidyl-tRNA hydrolase activity, to release the peptidyl moiety from tRNA, thereby preventing the accumulation of dissociated peptidyl-tRNA that could reduce the efficiency of translation. This protein also plays a role regulating cell survival and death. It promotes survival as part of an integrin-signaling pathway for cells attached to the extracellular matrix (ECM), but also promotes apoptosis in cells that have lost their attachment to the ECM, a process called anoikos. After loss of cell attachment to the ECM, this protein is phosphorylated, is released from the mitochondria into the cytosol, and promotes caspase-independent apoptosis through interactions with transcriptional regulators. This gene has been implicated in the development and progression of tumors, and mutations in this gene have been associated with an infantile multisystem neurologic, endocrine, and pancreatic disease (INMEPD) characterized by intellectual disability, postnatal microcephaly, progressive cerebellar atrophy, hearing impairment, polyneuropathy, failure to thrive, and organ fibrosis with exocrine pancreas insufficiency. Alternative splicing results in multiple transcript variants encoding different isoforms.Spécification
PTRH2 | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8R2Y8, Q9Y3E5 | |
BIT1, CGI 147, peptidyl tRNA hydrolase 2, PTH 2, PTH2, PTRH2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
217057, 287593, 51651 |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
PTRH2 | |
Liquid | |
Ptrh2 | |
PTRH2 Fusion Protein Ag0839 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Rat, Human, Mouse |