missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PVRL1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24713-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes an adhesion protein that plays a role in the organization of adherens junctions and tight junctions in epithelial and endothelial cells. The protein is a calcium(2+)-independent cell-cell adhesion molecule that belongs to the immunoglobulin superfamily and has 3 extracellular immunoglobulin-like loops, a single transmembrane domain, and a cytoplasmic region. This protein acts as a receptor for glycoprotein D of herpes simplex viruses 1 and 2, and pseudorabies virus and mediates viral entry into epithelial and neuronal cells. Mutations in this gene cause cleft lip and palate/ectodermal dysplasia 1 syndrome as well as non-syndromic cleft lip with or without cleft palate. Alternative splicing results in multiple transcript variants encoding proteins with distinct C-termini.Spécification
PVRL1 | |
Polyclonal | |
Unconjugated | |
NECTIN1 | |
CD111, CLPED1, ED4, Herpes virus entry mediator C, Herpesvirus entry mediator C, Herpesvirus Ig like receptor, HIgR, HVEC, nectin 1, OFC7, PRR, PRR1, PVRL1, PVRR, PVRR1, SK 12 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5818, 58235 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15223, Q9JKF6 | |
NECTIN1 | |
PVRL1 Fusion Protein Ag20074 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |