missing translation for 'onlineSavingsMsg'
Learn More

PVRL4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 21903-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16801455

  • 329.85€ / 150µL

Expédition estimée
08-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Poliovirus receptor-like proteins (PVRLs), such as PVRL4, are adhesion receptors of the immunoglobulin superfamily and function in cell-cell adhesion (Reymond et al., 2001 ).
Spécification

Spécification

PVRL4
0.41 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8R007, Q96NY8
Ig superfamily receptor LNIR, LNIR, nectin 4, poliovirus receptor related 4, PRR4, PVRL4
Rabbit
IgG
150 μL
-20°C
Primary
71740, 81607
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
NECTIN4
Liquid
Nectin4
PVRL4 Fusion Protein Ag14127
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Vidéos
FDS
Documents