missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PXMP3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22163-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes an integral peroxisomal membrane protein required for peroxisome biogenesis. The protein is thought to be involved in peroxisomal matrix protein import. Mutations in this gene result in one form of Zellweger syndrome and infantile Refsum disease. Alternative splicing results in multiple transcript variants encoding the same protein.Spécification
PXMP3 | |
Polyclonal | |
Unconjugated | |
PEX2 | |
PAF 1, PAF1, Peroxin 2, Peroxisomal membrane protein 3, Peroxisome assembly factor 1, Peroxisome biogenesis factor 2, PEX2, PMP3, PMP35, PXMP3, RING finger protein 72, RNF72 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19302, 5828 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P28328, P55098 | |
PEX2 | |
PXMP3 Fusion Protein Ag17815 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |