missing translation for 'onlineSavingsMsg'
Learn More

(R,R)-DIPAMP, 97 %, Thermo Scientific Chemicals

Catalog Number p-7030393
Click to view available options
Quantité:
100 mg
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
55739-58-7
C28H28O2P2
458.48
MFCD05863546
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
10884975
COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
55739-58-7
C28H28O2P2
458.48
MFCD05863546
QKZWXPLBVCKXNQ-UHFFFAOYNA-N
r,r-dipamp, 1,2-bis r-2-methoxyphenyl phenyl phosphino ethane, unii-blj831owlw, --dipamp, r,r-ethylenebis 2-methoxyphenyl phenylphosphine, r,r-1,2-bis 2-methoxyphenyl phenylphosphino ethane, blj831owlw, s,s-dipamp, 1r,2r---bis 2-methoxyphenyl phenylphosphino ethane
10884975
COC1=C(C=CC=C1)P(CCP(C1=CC=CC=C1)C1=C(OC)C=CC=C1)C1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau (R,R)-DIPAMP
Point de fusion 101°C to 105°C
Quantité 100 mg
Rotation optique −83° (c=1 in Chloroform)
Nom IUPAC (R)-(2-méthoxyphényl)-[2-[(2-méthoxyphényl)-phénylphosphanyl]éthyl]-phénylphosphane
Poids de la formule 458.47
Pourcentage de pureté 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'