missing translation for 'onlineSavingsMsg'
Learn More

RAB3GAP2 Mouse anti-Human, Clone: 3A4A1, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66648-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16818725

  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The protein encoded by this gene belongs to the RAB3 protein family, members of which are involved in regulated exocytosis of neurotransmitters and hormones. This protein forms the Rab3 GTPase-activating complex with RAB3GAP1, where it constitutes the regulatory subunit, whereas the latter functions as the catalytic subunit. This gene has the highest level of expression in the brain, consistent with it having a key role in neurodevelopment. Mutations in this gene are associated with Martsolf syndrome.
Spécification

Spécification

RAB3GAP2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H2M9
RAB3GAP2
RAB3GAP2 Fusion Protein Ag20254
150 μL
Primary
Human
Antibody
IgG2a
Western Blot
3A4A1
Unconjugated
RAB3GAP2
DKFZp434D245, FLJ14579, KIAA0839, p150, Rab3 GAP p150, Rab3 GAP regulatory subunit, RAB3 GAP150, RAB3GAP150, RAB3GAP2, RGAP iso
Mouse
Protein A
RUO
25782
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions