missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Rabbit OX40 / TNFRSF4 / CD134 Protein, Fc Tag

Marque:  ACROBiosystems OX0-R5254-1MG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877407

  • 1866.26€ / 1mg
Date d’expédition estimée: 07-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.3 kDa. As a result of different glycosylation, the protein migrates as 48 kDa and 68 kDa under reducing (R) condition, and 120-135 kDa under non-reducing (NR
Spécification

Spécification

Less than 1.0 EU / µg by the LAL method.
Immune Checkpoint Proteins
Powder
>85% as determined by SDS-PAGE
Rabbit O x 40, Fc Tag (O x 0-R5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 208 (Accession # O02764).
-20°C
OX40
Fc Tag
HEK293
Rabbit
47.3 kDa
1 mg
Ala 19 - Gln 208
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L
Unconjugated
Tris with Glycine, Arginine and NaCl, pH7.5
Vidéos
FDS
Documents
Certificats
Promotions

Promotions