missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Rabbit OX40 / TNFRSF4 / CD134 Protein, Fc Tag
387.09€ - 2262.06€
Spécification
Poids moléculaire | 47.3 kDa |
---|---|
Source | Rabbit O x 40, Fc Tag (O x 0-R5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 208 (Accession # O02764). |
Immunogène | Ala 19 - Gln 208 |
Conditions de stockage | -20°C |
Alias de gène | TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15877407
|
ACROBiosystems
OX0-R5254-1MG |
1 mg |
2262.06€
1mg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867407
|
ACROBiosystems
OX0-R5254-100UG |
100 μg |
387.09€
100µg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 47.3 kDa. As a result of different glycosylation, the protein migrates as 48 kDa and 68 kDa under reducing (R) condition, and 120-135 kDa under non-reducing (NRSpécification
47.3 kDa | |
Ala 19 - Gln 208 | |
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L | |
Unconjugated | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |
Rabbit O x 40, Fc Tag (O x 0-R5254) is expressed from human 293 cells (HEK293). It contains AA Ala 19 - Gln 208 (Accession # O02764). | |
-20°C | |
OX40 | |
Rabbit | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
Immune Checkpoint Proteins |