missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RABEP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14350-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
RABEP1 gene ontology annotations related to this gene include endocytosis.Spécification
RABEP1 | |
0.34 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35550, O35551, Q15276 | |
RAB5EP, Rabaptin 4, Rabaptin 5, Rabaptin 5alpha, RABEP1, RABPT5, RABPT5A | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
54189, 54190, 9135 |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
RABEP1 | |
Liquid | |
Rabep1 | |
RABEP1 Fusion Protein Ag5693 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |