missing translation for 'onlineSavingsMsg'
Learn More

RAC1/2/3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 16117-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16835624

  • 121.38€ / 20µL
Expédition estimée: 30-09-2022
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody can recognize Rac1, Rac2, and Rac3.

The protein encoded by this gene is a GTPase which belongs to the RAS superfamily of small GTP-binding proteins. Members of this superfamily appear to regulate a diverse array of cellular events, including the control of cell growth, cytoskeletal reorganization, and the activation of protein kinases. Two transcript variants encoding different isoforms have been found for this gene.
Spécification

Spécification

RAC1/2/3
Polyclonal
Unconjugated
RAC1
p21 Rac3, RAC3
Rabbit
Antigen Affinity Chromatography
RUO
170758, 19353, 19354, 363875, 366957, 5879, 5880, 5881, 688319
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P15153, P60763, P60764, P63000, P63001, Q05144, Q6RUV5
Rac1, Rac2, Rac3
RAC1/2/3 Fusion Protein Ag9045
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Vidéos
FDS
Documents
Certificats
Promotions

Promotions