missing translation for 'onlineSavingsMsg'
Learn More

RAD51 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14961-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16803904

  • 140.00€ / 20µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

RAD51 plays a critical role in homologous strand exchange, a key step in DNA repair through homologous recobination. It binds to single and double-stranded DNA and exhibits DNA-dependent ATPase activity. RAD51 catalyzes the recognition of homology and strand exchange between homologous DNA partners to form a joint molecule between a processed DNA break and the repair template. It binds to a single-stranded DNA in an ATP-dependent manner to form nucleoprotein filaments which are essential for the homology search and strand exchange. Mutations in the gene can results in breast cancer, mirror movements 2 and fanconi anemia.
Spécification

Spécification

RAD51
Polyclonal
Unconjugated
RAD51
BRCC5, HRAD51, HsRad51, HsT16930, RAD51, RAD51 homolog A, RAD51A, RECA
Rabbit
Antigen Affinity Chromatography
RUO
19361, 5888
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q06609, Q08297
RAD51
RAD51 Fusion Protein Ag6823
20 μL
Primary
Mouse, Human
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions