missing translation for 'onlineSavingsMsg'
Learn More

Radixin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13790-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802184

  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Ezrin, radixin and moesin (ERM) proteins are widely distributed proteins located in the cellular cortex, in microvilli and adherens junctions. They are mediate linkage of actin cytoskeleton to plasma membrane in many cells. ERM proteins contain an N-terminal FERM (4.1/ezrin/radixin/moesin) domain that binds to phosphatidylinositol-(4, 5)phosphate and cellular membrane proteins. The ERM, particularly ezrin, is important for reconstructing cell-surface architecture during T cell activation. Despite the high degree of homology, the three proteins exhibit a distinct receptor-specific pattern of phosphorylation. ERM activity is regulated in part by phosphorylation at a C-terminal threonine (ezrin-Thr567, radixin-Thr564, moesin-Thr558).
Spécification

Spécification

Radixin
Polyclonal
Unconjugated
RDX
DFNB24, radixin, RDX
Rabbit
Antigen Affinity Chromatography
RUO
19684, 315655, 5962
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P26043, P35241
Rdx
Radixin Fusion Protein Ag4813
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions