missing translation for 'onlineSavingsMsg'
Learn More

RAP1GAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14229-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802774

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

Rap1GAP specifically stimulates GTP hydrolytic activity of the monomeric G protein Rap1. Physical interaction between G alpha z, a member of the Gi family of trimeric G proteins, and Rap1GAP blocks the ability of regulators of G protein signaling to stimulate GTP hydrolysis of the alpha subunit, and also attenuates the ability of activated G alpha z to inhibit adenylyl cyclase. Rap1GAP is expressed in the brain, kidney and pancreas and may act as a signal integrator to coordinate and/or integrate Gz signaling and Rap1 signaling in cells. A novel isoform of Rapl GTPase-activating protein, designated Rap1GAPII, binds specifically to G alpha z. Stimulation of the Gi-coupled M2 Muscarinic receptor translocates Rap1GAPII from the cytosol to the membrane and decreases the amount of GTP-bound Rap1, resulting in the activation of ERK/MAPK.
Spécification

Spécification

RAP1GAP
Polyclonal
Unconjugated
RAP1GAP
KIAA0474, RAP1 GTPase activating protein, RAP1GA1, RAP1GAP, RAP1GAP1, RAP1GAPII, RAPGAP
Rabbit
Antigen Affinity Chromatography
RUO
110351, 313644, 5909
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A2ALS5, P47736
Rap1gap
RAP1GAP Fusion Protein Ag5469
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions