Learn More
ACROBiosystems Rat 4-1BB / TNFRSF9 Protein, His Tag (MALS verified)
423.38€ - 3108.82€
Spécification
Poids moléculaire | 19.5 kDa |
---|---|
Source | Rat 4-1 BB, His Tag (41 B-R52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 24 - Gln 187 (Accession # Q4W8J3-1). |
Immunogène | Thr 24 - Gln 187 |
Conditions de stockage | -20°C |
Alias de gène | TNFRSF9,4-1 BB,CD137,CDw137,ILA |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17256030
|
ACROBiosystems
41B-R52H3-1MG |
1 mg |
3108.82€
1mg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17246030
|
ACROBiosystems
41B-R52H3-100UG |
100 μg |
423.38€
100µg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
4-1BB is also known as CD137, tumor necrosis factor receptor superfamily member 9 (TNFRSF9), induced by lymphocyte activation (ILA), is a co-stimulatory molecule of the tumor necrosis factor (TNF) receptor superfamily. CD137 can be expressed by activated T cells, but to a larger extent on CD8 than on CD4 T cells. In addition, CD137 expression is found on dendritic cells, follicular dendritic cells, natural killer cells, granulocytes and cells of blood vessel walls at sites of inflammation. The best characterized activity of CD137 is its costimulatory activity for activated T cells. Crosslinking of CD137 enhances T cell proliferation, IL-2 secretion survival and cytolytic activity. Further, it can enhance immune activity to eliminate tumors in mice. CD137 can enhance activation-induced T cell apoptosis when triggered by engagement of the TCR/CD3 complex. In addition, 4-1BB/4-1BBL co-stimulatory pathway has been shown to augment secondary CTL responses to several viruses, and meanwhile augment anti-tumor immunity. 4-1BB thus is a promising candidate for immunotherapy of human cancer. CD137 has been shown to interact with TRAF2.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 19.5 kDa. The protein migrates as 28-38 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
19.5 kDa | |
Thr 24 - Gln 187 | |
TNFRSF9,4-1 BB,CD137,CDw137,ILA | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
Rat 4-1 BB, His Tag (41 B-R52H3) is expressed from human 293 cells (HEK293). It contains AA Thr 24 - Gln 187 (Accession # Q4W8J3-1). | |
-20°C | |
4-1 BB | |
Rat | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |