Learn More
RBM15B Mouse anti-Human, Unconjugated, Clone: 1C2C11, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67506-1-IG-150UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16830176
  • 399.00€ / 150µL
Expédition estimée: 03-05-2024
Description
The RBM15B antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RBM15B. This antibody recognizes human antigen. The RBM15B antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
RBM15B
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
1C2C11
Unconjugated
150 μL
29890
IgG1