missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15374-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes an RNA binding protein. The encoded protein is part of the spliceosome complex and functions in the second catalytic step of mRNA splicing. Alternatively spliced transcript variants have been described. Related pseudogenes exist on chromosomes 9 and 15.Spécification
RBM17 | |
Polyclonal | |
Unconjugated | |
RBM17 | |
45 kDa splicing factor, RBM17, RNA binding motif protein 17, SPF45, Splicing factor 45 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
291295, 76938, 84991 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8JZX4, Q96I25 | |
Rbm17 | |
RBM17 Fusion Protein Ag0985 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |