missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM47 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23902-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The function of this protein remains unknown.Spécification
RBM47 | |
Polyclonal | |
Unconjugated | |
RBM47 | |
NET18, RBM47, RNA binding motif protein 47, RNA binding protein 47 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305340, 54502 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A0AV96, Q66H68 | |
Rbm47 | |
RBM47 Fusion Protein Ag20984 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |