Learn More
RBMX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA555356
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: NYRAPKDSEE IDDVTRQLQE KGCGARTPSP SLSESSEDEK PTKKHKKDKK EKKKKKKEKE KADREVQAEQ PSSSSPRRKT VKEKDDTGPK KHSSKNSERA QKSEPREGQK LPKSR Highest antigen sequence identity to the following orthologs: Mouse - 64%, Rat - 68%.
RBMX2 Polyclonal antibody specifically detects RBMX2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spécification
RBMX2 | |
Polyclonal | |
Unconjugated | |
RBMX2 | |
2700089A17Rik; 2810411A03Rik; CGI-79; Rbmx2; RGD1562693; RNA binding motif protein, X-linked 2; RNA-binding motif protein, X-linked 2; Snu17 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51634 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y388 | |
RBMX2 | |
Recombinant protein corresponding to Human RBMX2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only