Learn More
RBP7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 14541-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
 Afficher plus de versions de ce produit
Code produit. 16823234
  • 399.00€ / 150µL
Expédition estimée: 15-04-2024
pour voir le stock disponible 
Description
Description
The RBP7 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RBP7. This antibody recognizes human, mouse, rat antigen. The RBP7 antibody has been validated for the following applications: IF, IHC, IP, WB, ELISA analysis.
Spécification
Spécification
RBP7
Polyclonal
Unconjugated
150 μL
116362
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
233 μg/mL
Rabbit
Primary
Human, Rat, Mouse