missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCC2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 16755-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The protein encoded by this gene is a member of the RecQ DEAH helicase family and interacts with the BRCT repeats of breast cancer, type 1 (BRCA1). The bound complex is important in the normal double-strand break repair function of breast cancer, type 1 (BRCA1). This gene may be a target of germline cancer-inducing mutations.Spécification
RCC2 | |
Polyclonal | |
Unconjugated | |
RCC2 | |
KIAA1470, Protein RCC2, RCC1 like protein TD 60, RCC2, TD 60, TD60 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55920 | |
-20°C | |
Liquid |
Western Blot | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9P258 | |
RCC2 | |
RCC2 Fusion Protein Ag10271 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |