missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ REC14 Recombinant Protein
Marque: Abnova™ H00080349-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Spécification
AAH10080 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
59.29 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MTNQYGILFKQEQAHDDAIWSVAWGTNKKENSETVVTGSLDDLVKVWKWRDERLDLQWSLEGHQLGVVSVDISHTLPIAASSSLDAHIRLWDLENGKQIKSIDAGPVDAWTLAFSPDSQYLATGTHVGKVNIFGVESGKKEYSLDTRGKFILSIAYSPDGKYLASGAIDGIINIFDIATGKLLHTLEGHAMPIRSLTFSPDSQLLVTASDDGYIKIYDVQHANLAGTLSGHASWVLNVAFCPDDTHFVSSSSDKSVKVWDVGTRTCVHTFFDHQDQVWGVKYNGNGSKIVSVGDDQEIHIYDCPI | |
REC14/SKI8 | |
WDR61 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, ELISA, Protein Array, Western Blot | |
80349 | |
REC14 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WDR61 | |
Human | |
Recombinant | |
Solution |