missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant C. trachomatis Large cysteine-rich periplasmic protein OmcB Protein
A DNA sequence encoding the Chlamydia trachomatis (strain D/UW-3/Cx) Large cysteine-rich periplasmic protein OmcB, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7015-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_219955.1 | |
Large cysteine-rich periplasmic protein OmcB Protein | |
Research Use Only | |
omcB | |
Recombinant Protein | |
E. coli | |
LADTKAKDNT SHKSKKARKN HSKETPVDRK EVAPVHESKA TGPKQDSCFG RMYTVKVNDD RNVEITQAVP EYATVGSPYP IEITATGKRD CVDVIITQQL PCEAEFVRSD PATTPTADGK LVWKIDRLGQ GEKSKITVWV KPLKEGCCFT AATVCA | |
Greater than 90% as determined by SDS-PAGE. |
884223 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
C. trachomatis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |