missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Cat SDF-1 beta / CXCL12 Protein
Recombinant Protein
Marque: enQuireBio™ QP5218-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_001009847.1 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
CXCL12 | |
Cat | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
SDF-1 beta / CXCL12 Protein | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRF KM |