missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken GH Protein

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10632-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15974468

  • 203.00€ / 100µg
Expédition estimée: 24-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

GH Protein
Research Use Only
GH Chicken Recombinantis fully biologically active in homologous assays and in PDF-P1 3B9 cells stably transfected with rabbit GH receptors.
Chicken
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.3% NaHCO3 adjusted to pH 8.
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MAPGSWFSPL LIAVVTLGLP QEAAATFPAM PLSNLFANAV LRAQHLHLLA AETYKEFERT YIPEDQRYTN KNSQAAFCYS ETIPAPTGKD DAQQKSDMEL LRFSLVLIQS WLTPVQYLSK VFTNNLVFGT SDRVFEKLKD LEEGIQALMR ELEDRSPRGP QLLRPTYDKF DIHLRNEDAL LKNYGLLSCF KKDLHKVETY LKVMKCRRFG ESNCTI
Greater than 99.0% as determined by:(a) Analysis by SDS-PAGE gel. (b) Analysis by SEC-HPLC.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions