missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Envelope-2 & 4 Protein

A cDNA sequence encoding the Dengue Envelope-2 & 4 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11627-500ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15947848

  • 710.00€ / 500µg

Expédition estimée
20-11-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Spécification

Spécification

Dengue Envelope-2 & 4 Protein
500 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
The protein solution contains Phosphate buffered saline, pH 7.4.
Vidéos
Documents