Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 3 VLP Protein
Recombinant Protein
Marque:  enQuireBio™ QP10481-500ug
Code nomenclature Nacres: NA.28
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15983998
  • 5365.00€ / 500µg
Expédition estimée: 29-08-2024
pour voir le stock disponible 
Description
Description
This VLP protein is combinantion of the C, E1 and E2 sequences. Virus-Like Particles (VLPs) are a new form of protein expression that consist of protein shells with outer proteins that are specific to the given virus. VLPs are immunogenic but are non-infectious, lacking the primary genetic material of the virus. Additionally, VLPs more effectively activate aspects of the immune response, which improves immune stimulation and immunological memory. VLP-based vaccines have also shown to provide effective protection and are in current use for several diseases.
Spécification
Spécification
Serotype 3 VLP Protein
Research Use Only
Recombinant Protein
Human
10mM sodium phosphate, 20mM sodium citrate, 154mM sodium chloride, pH7.4.
500 μg
Untested
Dengue virus (Strain: Sri Lanka D3/H/IMTSSA-SRI/2000/1266)
Native
>95% as determined by SDS-PAGE.