missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Annexin A4 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Annexin A4, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5653-ec-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Annexin A4 Protein | |
Research Use Only | |
ANXA4 | |
Recombinant Protein | |
E. coli | |
AAKGGTVKPA SGFSATEDAQ TLRKAMKGLG TDEDAIISVL APRNTSQRQE IRTAYKSTIG RDLMDDLKSE LSGNFERVIV GMITPTVLYD VQELRRAMKG SGTDEGCLIE ILASRTPEEL RCINQTYQLQ YGRSLEDVIR SDTSFMFQRV LVSLSAGGRD EGNFLDDALM RQDAQDLYEA GEKKWGTDEV KFLTVLCSRN RNHLLHVFDE YKRISQKDIE QGIKSETSGS FEDALLAIVK CMRNKSAYFA ERLYKSMKGL GTDDNTLIRV MVSRAEIDMM DIRESFKRLY GKSLYSFIKG DTSGDYRKVL LILCGGDD | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Dog | |
His-SUMO | |
Tris-based buffer, 50% glycerol |