missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coi CYSH Protein
A cDNA sequence encoding the CYSH was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11585-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
E. Coi CYSH Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSKLDLNALN ELPKVDRILA LAETNAELEK LDAEGRVAWA LDNLPGEYVL SSSFGIQAAV SLHLVNQIRP DIPVILTDTG YLFPETYRFI DELTDKLKLN LKVYRATESA AWQEARYGKL WEQGVEGIEK YNDINKVEPM NRALKELNAQ TWFAGLRREQ SGSRANLPVL AIQRGVFKVL PIIDWDNRTI YQYLQKHGLK YHPLWDEGYL SVGDTHTTRK WEPGMAEEET RFFGLKRECG LHEG | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
CYSH protein solution (0.5 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 50mM NaCl. |