Learn More
enQuireBio™ Recombinant E. Coli Ecotin Protein
A cDNA sequence encoding the Ecotin was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11741-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
Ecotin Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
The Ecotin protein solution (1 mg/ml) is formulated in 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 50mM NaCl and 10% glycerol. |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAESVQPLEK IAPYPQAEKG MKRQVIQLTP QEDESTLKVE LLIGQTLEVD CNLHRLGGKL ENKTLEGWGY DYYVFDKVSS PVSTMMACPD GKKEKKFVTA YLGDAGMLRY NSKLPIVVYT PDNVDVKYRV WKAEEKIDNA VVR |