Learn More
enQuireBio™ Recombinant E. coli fkpA Protein
A DNA sequence encoding the Escherichia coli (strain K12) fkpA, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP7261-ec-200ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
WP_000838261.1 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
fkpA Protein | |
Research Use Only | |
fkpA | |
Recombinant Protein | |
E. coli | |
AEAAKPATTA DSKAAFKNDD QKSAYALGAS LGRYMENSLK EQEKLGIKLD KDQLIAGVQD AFADKSKLSD QEIEQTLQAF EARVKSSAQA KMEKDAADNE AKGKEYREKF AKEKGVKTSS TGLVYQVVEA GKGEAPKDSD TVVVNYKGTL IDGKEFDNSY TRGEPLSFRL DGVIPGWTEG LKNIKKGGKI KLVIPPELAY GKAGVPGIPP NSTLVFDVEL LDVKPAPKAD AKPEADAKAA DSAKK | |
Greater than 90% as determined by SDS-PAGE. |