missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. Coli UPP1 Protein

A cDNA sequence encoding the UPP1 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP13899-1mg

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975059

  • 3401.86€ / 1mg

Date d’expédition estimée: 25-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Spécification

UPP1 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
E. coli
His
UPP1 solution (1 mg/ml) contains 20 mM Tris-HCl buffer (pH 8.0), 1mM DTT, 50mM NaCl and 10% glycerol.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MSKSDVFHLG LTKNDLQGAT LAIVPGDPDR VEKIAALMDK PVKLASHREF TTWRAELDGK PVIVCSTGIG GPSTSIAVEE LAQLGIRTFL RIGTTGAIQP HINVGDVLVT TASVRLDGAS LHFAPLEFPA VADFECTTAL VEAAKSIGAT THVGVTASSD TFYPGQERYD TYSGRVVRHF KGSMEEWQAM GVMNYEMESA TLLTMCASQG LRAGMVAGVI VNRTQQEIPN AETMKQTESH AVKIVVEAAR RLL
Vidéos
FDS
Documents
Certificats