missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Ebola Virus Ebola Zaire GP Protein

A cDNA sequence encoding the Ebola Zaire GP was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP11727-100ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15968158

Quantité Prix
1 194.39€ / 100µg
6 182.42€ / 100µg
Date d’expédition estimée: 22-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

Ebola Zaire GP Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Ebola Virus
His
>95% pure as determined by 12% SDS-PAGE (Coomassie blue stain).
Research Use Only
Recombinant Protein
E. coli
Ebola Zaire GP protein solution is supplied in Phosphate buffer with 25mM arginine and 0.02% sodium azide.
Vidéos
FDS
Documents
Certificats