missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant HIV Type-O gp41 Protein
A cDNA sequence encoding the HIV Type-O gp41 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12238-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
HIV Type-O gp41 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25mM Tris-HCl pH 8.0 and 50% Glycerol. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HIV | |
Untagged | |
Greater than 90.0% as determined by SDS-PAGE. |