Learn More
enQuireBio™ Recombinant HSV HCV Genotype 1b Protein
A cDNA sequence encoding the HCV Genotype 1b was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP12159-100ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
HSV HCV Genotype 1b Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
HSV | |
Untagged |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Sterile filtered solution containing 1x PBS and 25mM Arginine. |