Learn More
enQuireBio™ Recombinant Human ABHD12B Protein
A cDNA sequence encoding the ABHD12B was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP10927-1mg
Informations supplémentaires : Poids : 0.01000kg
Spécification
ABHD12B Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ABHD12B protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol. |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMLGIWHT VPSCRGEDAK GKDCCWYEAA LRDGNPIIVY LHGSAEHRAA SHRLKLVKVL SDGGFHVLSV DYRGFGDSTG KPTEEGLTTD AICVYEWTKA RSGITPVCLW GHSLGTGVAT NAAKVLEEKG CPVDAIVLEA PFTNMWVASI NYPLLKIYRN IPGFLRTLMD ALRKDKIIFP NDENVKFLSS PLLILHGEDD RTVPLEYGKK LYEIARNAYR NKERVKMVIF PPGFQHNLLC KSPTLLITVR DFLSKQWS |