missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human AK5 Protein

A cDNA sequence encoding the AK5 was constructed and used to recombinantly synthesize the protein.

Marque:  enQuireBio™ QP10992-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15975798

Quantité Prix
1 171.64€ / 10µg
10 161.27€ / 10µg
Date d’expédition estimée: 18-11-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Spécification

Spécification

26289
Greater than 85% as determined by SDS-PAGE.
Research Use Only
AK5
Human
His
AK5 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol and 1mM DTT.
AK5 Protein
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMNTNDA KEYMARREIP QLFESLLNGL MCSKPEDPVE YLESCLQKVK ELGGCDKVKW DTFVSQEKKT LPPLNGGQSR RSFLRNVMPG NSNFPYRRYD RLPPIHQFSI ESDTDLSETA ELIEEYEVFD PTRPRPKIIL VIGGPGSGKG TQSLKIAERY GFQYISVGEL LRKKIHSTSS NRKWSLIAKI ITTGELAPQE TTITEIKQKL MQIPDEEGIV IDGFPRDVAQ ALSFEDQICT PDLVVFLACA NQRLKERLLK RAEQQGRPDD NVKATQRRLM NFKQNAAPLV KYFQEKGLIM TFDADRDEDE VFYDISMAVD NKLFPNKEAA AGSSDLDPSM ILDTGEIIDT GSDYEDQGDD QLNVFGEDTM GGFMEDLRKC KIIFIIGGPG SGKGTQCEKL VEKYGFTHLS TGELLREELA SESERSKLIR DIMERGDLVP SGIVLELLKE AMVASLGDTR GFLIDGYPRE VKQGEEFGRR IGDPQLVICM DCSADTMTNR LLQRSRSSLP VDDTTKTIAK RLEAYYRASI PVIAYYETKT QLHKINAEGT PEDVFLQLCT AIDSIF
Vidéos
FDS
Documents
Certificats