missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Alcohol dehydrogenase 1C Protein

A DNA sequence encoding the Homo sapiens (Human) Alcohol dehydrogenase 1C, was expressed in the hosts and tags indicated.

Marque:  enQuireBio™ QP5614-ec-10ug

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15906449

Cet article est actuellement indisponible ou discontinu. Veuillez consulter la page produit pour de possible alternative dans l’onglet


Description

Description

Spécification

Spécification

NP_000660.1
Alcohol dehydrogenase 1C Protein
Research Use Only
ADH1C
Recombinant Protein
E. coli
STAGKVIKCK AAVLWELKKP FSIEEVEVAP PKAHEVRIKM VAAGICRSDE HVVSGNLVTP LPVILGHEAA GIVESVGEGV TTVKPGDKVI PLFTPQCGKC RICKNPESNY CLKNDLGNPR GTLQDGTRRF TCSGKPIHHF VGVSTFSQYT VVDENAVAKI DAASPLEKVC LIGCGFSTGY GSAVKVAKVT PGSTCAVFGL GGVGLSVVMG CKAAGAARII AVDINKDKFA KAKELGATEC INPQDYKKPI QEVLKEMTDG GVDFSFEVIG RLDTMMASLL CCHEACGTSV IVGVPPDSQN LSINPMLLLT GRTWKGAIFG GFKSKESVPK LVADFMAKKF SLDALITNIL PFEKINEGFD LLRSGKSIRT VLTF
Greater than 80% as determined by SDS-PAGE.
126
10 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions