missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Alcohol dehydrogenase 1C Protein
A DNA sequence encoding the Homo sapiens (Human) Alcohol dehydrogenase 1C, was expressed in the hosts and tags indicated.
Marque: enQuireBio™ QP5614-ec-500ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
NP_000660.1 | |
Alcohol dehydrogenase 1C Protein | |
Research Use Only | |
ADH1C | |
Recombinant Protein | |
E. coli | |
STAGKVIKCK AAVLWELKKP FSIEEVEVAP PKAHEVRIKM VAAGICRSDE HVVSGNLVTP LPVILGHEAA GIVESVGEGV TTVKPGDKVI PLFTPQCGKC RICKNPESNY CLKNDLGNPR GTLQDGTRRF TCSGKPIHHF VGVSTFSQYT VVDENAVAKI DAASPLEKVC LIGCGFSTGY GSAVKVAKVT PGSTCAVFGL GGVGLSVVMG CKAAGAARII AVDINKDKFA KAKELGATEC INPQDYKKPI QEVLKEMTDG GVDFSFEVIG RLDTMMASLL CCHEACGTSV IVGVPPDSQN LSINPMLLLT GRTWKGAIFG GFKSKESVPK LVADFMAKKF SLDALITNIL PFEKINEGFD LLRSGKSIRT VLTF | |
Greater than 80% as determined by SDS-PAGE. |
126 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |