missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANGPTL3 Protein
A cDNA sequence encoding the ANGPTL3 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11020-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
27329 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPSRIDQDN SSFDSLSPEP KSRFAMLDDV KILANGLLQL GHGLKDFVHK TKGQINDIFQ KLNIFDQSFY DLSLQTSEIK EEEKELRRTT YKLQVKNEEV KNMSLELNSK LESLLEEKIL LQQKVKYLEE QLTNLIQNQP ETPEHPEVTS LKTFVEKQDN SIKDLLQTVE DQYKQLNQQH SQIKEIENQL RRTSIQEPTE ISLSSKPRAP RTTPFLQLNE IRNVKHDGIP AECTTIYNRG EHTSGMYAIR PSNSQVFHVY CDVISGSPWT LIQHRIDGSQ NFNETWENYK YGFGRLDGEF WLGLEKIYSI VKQSNYVLRI ELEDWKDNKH YIEYSFYLGN HETNYTLHLV AITGNVPNAI PENKDLVFST WDHKAKGHFN CPEGYSGGWW WHDECGENNL NGKYNKPRAK SKPERRRGLS WKSQNGRLYS IKSTKMLIHP TDSESFEHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |
ANGPTL3 Protein | |
Research Use Only | |
ANGPTL3 | |
Human | |
His | |
ANGPTL3 protein solution (0.25 mg/ml) contains Buffered Saline (pH 7.4) and 10% glycerol. |