missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ANGPTL6 Protein
A cDNA sequence encoding the ANGPTL6 was constructed and used to recombinantly synthesize the protein.
Marque: enQuireBio™ QP11021-10ug
Informations supplémentaires : Poids : 0.01000kg
Spécification
83854 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS RAGAPRCTYT FVLPPQKFTG AVCWSGPAST RATPEAANAS ELAALRMRVG RHEELLRELQ RLAAADGAVA GEVRALRKES RGLSARLGQL RAQLQHEAGP GAGPGADLGA EPAAALALLG ERVLNASAEA QRAAARFHQL DVKFRELAQL VTQQSSLIAR LERLCPGGAG GQQQVLPPPP LVPVVPVRLV GSTSDTSRML DPAPEPQRDQ TQRQQEPMAS PMPAGHPAVP TKPVGPWQDC AEARQAGHEQ SGVYELRVGR HVVSVWCEQQ LEGGGWTVIQ RRQDGSVNFF TTWQHYKAGF GRPDGEYWLG LEPVYQLTSR GDHELLVLLE DWGGRGARAH YDGFSLEPES DHYRLRLGQY HGDAGDSLSW HNDKPFSTVD RDRDSYSGNC ALYQRGGWWY HACAHSNLNG VWHHGGHYRS RYQDGVYWAE FRGGAYSLRK AAMLIRPLKL | |
Greater than 90.0% as determined by SDS-PAGE. |
ANGPTL6 Protein | |
Research Use Only | |
ANGPTL6 | |
Human | |
His | |
Filtered (0.4 microm) and lyophilized from a 0.5 mg/ml solution containing 0.05M Acetate buffer pH 4.0. |